missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human APOA1 Protein

A cDNA sequence encoding the APOA1 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11043-100ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15975958

Quantité Prix
1 171.64€ / 100µg
10 161.27€ / 100µg
Date d’expédition estimée: 25-11-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!



Description

Description

Spécification

Spécification

335
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
APOA1
Human
Untagged
The APOA1 1 mg/ml solution contains 10mM Ammonium Bicarbonate at pH 7.4.
APOA1 Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Native Protein
MKAAVLTLAV LFLTGSQARH FWQQDEPPQS PWDRVKDLAT VYVDVLKDSG RDYVSQFEGS ALGKQLNLKL LDNWDSVTST FSKLREQLGP VTQEFWDNLE KETEGLRQEM SKDLEEVKAK VQPYLDDFQK KWQEEMELYR QKVEPLRAEL QEGARQKLHE LQEKLSPLGE EMRDRARAHV DALRTHLAPY SDELRQRLAA RLEALKENGG ARLAEYHAKA TEHLSTLSEK AKPALEDLRQ GLLPVLESFK VSFLSALEEY TKKLNTQ
Vidéos
FDS
Documents
Certificats