missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ASMT Protein

A cDNA sequence encoding the ASMT was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11087-20ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15976098

  • 203.00€ / 20µg
Expédition estimée: 10-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

ASMT Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSSEDQAYR LLNDYANGFM VSQVLFAACE LGVFDLLAEA PGPLDVAAVA AGVRASAHGT ELLLDICVSL KLLKVETRGG KAFYRNTELS SDYLTTVSPT SQCSMLKYMG RTSYRCWGHL ADAVREGRNQ YLETFGVPAE ELFTAIYRSE GERLQFMQAL QEVWSVNGRS VLTAFDLSVF PLMCDLGGDF FKDPLPEADL YILARVLHDW ADGKCSHLLE RIYHTCKPGG GILVIESLLD EDRRGPLLTQ LYSLNMLVQT EGQERTPTHY HMLLSSAGFR DFQFKKTGAI YDAILARK
Greater than 90.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The ASMT solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) , 1M Urea and 10% glycerol.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions