missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BDH1, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) BDH1, mitochondrial, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5713-ec-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_004042.1 | |
BDH1, mitochondrial Protein | |
Research Use Only | |
BDH1 | |
Recombinant Protein | |
E. coli | |
ASAAEPVGSK AVLVTGCDSG FGFSLAKHLH SKGFLVFAGC LMKDKGHDGV KELDSLNSDR LRTVQLNVCS SEEVEKVVEI VRSSLKDPEK GMWGLVNNAG ISTFGEVEFT SLETYKQVAE VNLWGTVRMT KSFLPLIRRA KGRVVNISSM LGRMANPARS PYCITKFGVE AFSDCLRYEM YPLGVKVSVV EPGNFIAATS LYSPESIQAI AKKMWEELPE VVRKDYGKKY FDEKIAKMET YCSSGSTDTS PVIDAVTHAL TATTPYTRYH PMDYYWWLRM QIMTHLPGAI SDMIYIR | |
Greater than 90% as determined by SDS-PAGE. |
622 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |