missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BIN1 Protein

A cDNA sequence encoding the BIN1 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11162-20ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15986338

  • 203.00€ / 20µg
Expédition estimée: 17-01-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

BIN1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MAEMGSKGVT AGKIASNVQK KLTRAQEKVL QKLGKADETK DEQFEQCVQN FNKQLTEGTR LQKDLRTYLA SVKAMHEASK KLNECLQEVY EPDWPGRDEA NKIAENNDLL WMDYHQKLVD QALLTMDTYL GQFPDIKSRI AKRGRKLVDY DSARHHYESL QTAKKKDEAK IAKAEEELIK AQKVFEEMNV DLQEELPSLW NSRVGFYVNT FQSIAGLEEN FHKEMSKLNQ NLNDVLVGLE KQHGSNTFTV KAQPSDNAPA KGNKSPSPPD GSPAATPEIR VNHEPEPAGG ATPGATLPKS PSQPAEASEV AGGTQPAAGA QEPGETAASE AASSSLPAVV VETFPATVNG TVEGGSGAGR LDLPPGFMFK VQAQHDYTAT DTDELQLRAG DVVLVIPFQN PEEQDEGWLM GVKESDWNQH KELEKCRGVF PENFTERVP
Greater than 90.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
BIN1 protein solution (1 mg/ml) containing 20mM Tris buffer pH 8.0, 1mM DTT and 10% glycerol.
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions