missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Breast carcinoma-amplified sequence 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Breast carcinoma-amplified sequence 1, was expressed in the hosts and tags indicated.

Marque:  enQuireBio™ QP5705-ye-100ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15947149

  • 615.00€ / 100µg
Expédition estimée: 24-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

NP_003648.2
Breast carcinoma-amplified sequence 1 Protein
Research Use Only
BCAS1
Recombinant Protein
Yeast
MGNQMSVPQR VEDQENEPEA ETYQDNASAL NGVPVVVSTH TVQHLEEVDL GISVKTDNVA TSSPETTEIS AVADANGKNL GKEAKPEAPA AKSRFFLMLS RPVPGRTGDQ AADSSLGSVK LDVSSNKAPA NKDPSESWTL PVAAGPGQDT DKTPGHAPAQ DKVLSAARDP TLLPPETGGA GGEAPSKPKD SSFFDKFFKL DKGQEKVPGD SQQEAKRAEH QDKVDEVPGL SGQSDDVPAG KDIVDGKEKE GQELGTADCS VPGDPEGLET AKDDSQAAAI AENNNSIMSF FKTLVSPNKA ETKKDPEDTG AEKSPTTSAD LKSDKANFTS QETQGAGKNS KGCNPSGHTQ SVTTPEPAKE GTKEKSGPTS LPLGKLFWKK SVKEDSVPTG AEENVVCESP VEIIKSKEVE SALQTVDLNE GDAAPEPTEA KLKREESKPR TSLMAFLRQM SVKGDGGITH SEEINGKDSS CQTSDSTEKT ITPPEPEPTG APQKGKEGSS KDKKSAAEMN KQKSNKQEAK EPAQCTEQAT VDTNSLQNGD KLQKRPEKRQ QSLGGFFKGL GPKRMLDAQV QTDPVSIGPV GKSK
Greater than 90% as determined by SDS-PAGE.
8537
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions