missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BUB3 Protein
A cDNA sequence encoding the BUB3 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11212-20ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
BUB3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMTGSNEF KLNQPPEDGI SSVKFSPNTS QFLLVSSWDT SVRLYDVPAN SMRLKYQHTG AVLDCAFYDP THAWSGGLDH QLKMHDLNTD QENLVGTHDA PIRCVEYCPE VNVMVTGSWD QTVKLWDPRT PCNAGTFSQP EKVYTLSVSG DRLIVGTAGR RVLVWDLRNM GYVQQRRESS LKYQTRCIRA FPNKQGYVLS SIEGRVAVEY LDPSPEVQKK KYAFKCHRLK ENNIEQIYPV NAISFHNIHN TFATGGSDGF VNIWDPFNKK RLCQFHRYPT SIASLAFSND GTTLAIASSY MYEMDDTEHP EDGIFIRQVT DAETKPKSPC T | |
Greater than 85.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
BUB3 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |