Learn More
enQuireBio™ Recombinant Human Caspase-14 / CASP14 Protein
A DNA sequence encoding the Homo sapiens (Human) Caspase-14 / CASP14, was expressed in the hosts and tags indicated.
Marque:  enQuireBio™ QP5761-ec-500ug
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15997559
  • 1440.00€ / 500µg
Expédition estimée: 10-05-2024
Spécification
NP_036246.1
Caspase-14 / CASP14 Protein
Research Use Only
CASP14
Recombinant Protein
E. coli
KDSPQTIPTY TDALHVYSTV EGYIAYRHDQ KGSCFIQTLV DVFTKRKGHI LELLTEVTRR MAEAELVQEG KARKTNPEIQ STLRKRLY
Greater than 90% as determined by SDS-PAGE.
23581
500 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol