missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL20 / MIP-3 alpha Protein
Recombinant Protein
Marque: enQuireBio™ QP5271-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001123518.1 | |
CCL20 / MIP-3 alpha Protein | |
Research Use Only | |
CCL20 | |
Recombinant Protein | |
E. coli | |
ASNFDCCLGY TDRILHPKFI VGFTRQLANE GCDINAIIFH TKKKLSVCAN PKQTWVKYIV RLLSKKVKNM | |
Greater than 95% as determined by SDS-PAGE. |
6364 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Human CCL20 / MIP-3 alpha induces Primary human T cell chemotaxis. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |