missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD99 Protein
A cDNA sequence encoding the CD99 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11359-HIS-2ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
4267 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MARGAALALL LFGLLGVLVA APDGGFDLSD ALPDNENKKP TAIPKKPSAG DDFDLGDAVV DGENDDPRPP NPPKPMPNPN PNHPSSSGSF SDADLADGVS GGEGKGGSDG GGSHRKEGEE ADAPGVIPGI VGAVVVAVAG AISSFIAYQK KKLCFKENAE QGEVDMESHR NANAEPAVQR TLLEK |
CD99 Protein | |
Research Use Only | |
CD99 | |
Human | |
His | |
CD99 at 0.1 mg/ml in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol. |