missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CDK16 Protein

A cDNA sequence encoding the CDK16 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11373-5ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15997058

  • 131.00€ / 5µg
Expédition estimée: 10-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

CDK16 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSGFGKLET YIKLDKLGEG TYATVYKGKS KLTDNLVALK EIRLEHEEGA PCTAIREVSL LKDLKHANIV TLHDIIHTEK SLTLVFEYLD KDLKQYLDDC GNIINMHNVK LFLFQLLRGL AYCHRQKVLH RDLKPQNLLI NERGELKLAD FGLARAKSIP TKTYSNEVVT LWYRPPDILL GSTDYSTQID MWGVGCIFYE MATGRPLFPG STVEEQLHFI FRILGTPTEE TWPGILSNEE FKTYNYPKYR AEALLSHAPR LDSDGADLLT KLLQFEGRNR ISAEDAMKHP FFLSLGERIH KLPDTTSIFA LKEIQLQKEA SLRSSSMPDS GRPAFRVVDT EF
Greater than 95% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
CDK16 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl and 10% glycerol.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions