missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CLPP Protein

A cDNA sequence encoding the CLPP was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11451-2ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15947308

  • 107.00€ / 2µg

Expédition estimée
23-11-2020

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Spécification

Spécification

CLPP Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
CLPP solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 7.5), 2mM DTT, 20% glycerol and 100mM NaCl.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MPLIPIVVEQ TGRGERAYDI YSRLLRERIV CVMGPIDDSV ASLVIAQLLF LQSESNKKPI HMYINSPGGV VTAGLAIYDT MQYILNPICT WCVGQAASMG SLLLAAGTPG MRHSLPNSRI MIHQPSGGAR GQATDIAIQA EEIMKLKKQL YNIYAKHTKQ SLQVIESAME RDRYMSPMEA QEFGILDKVL VHPPQDGEDE PTLVQKEPVE AAPAAEPVPA ST
Vidéos
Documents