missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CRYZ Protein

A cDNA sequence encoding the CRYZ was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11525-5ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15967528

Quantité Prix
1 114.77€ / 5µg
10 107.33€ / 5µg
Date d’expédition estimée: 30-11-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!



Description

Description

Spécification

Spécification

CRYZ Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
CRYZ protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMATGQKL MRAVRVFEFG GPEVLKLRSD IAVPIPKDHQ VLIKVHACGV NPVETYIRSG TYSRKPLLPY TPGSDVAGVI EAVGDNASAF KKGDRVFTSS TISGGYAEYA LAADHTVYKL PEKLDFKQGA AIGIPYFTAY RALIHSACVK AGESVLVHGA SGGVGLAACQ IARAYGLKIL GTAGTEEGQK IVLQNGAHEV FNHREVNYID KIKKYVGEKG IDIIIEMLAN VNLSKDLSLL SHGGRVIVVG SRGTIEINPR DTMAKESSII GVTLFSSTKE EFQQYAAALQ AGMEIGWLKP VIGSQYPLEK VAEAHENIIH GSGATGKMIL LL
Vidéos
FDS
Documents
Certificats