Learn More
enQuireBio™ Recombinant Human CSN2 Protein
A cDNA sequence encoding the CSN2 was constructed and used to recombinantly synthesize the protein.
Marque:  enQuireBio™ QP11531-10ug
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15947548
  • 203.00€ / 10µg
Expédition estimée: 10-05-2024
Spécification
CSN2 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMRETIE SLSSSEESIT EYKQKVEKVK HEDQQQGEDE HQDKIYPSFQ PQPLIYPFVE PIPYGFLPQN ILPLAQPAVV LPVPQPEIME VPKAKDTVYT KGRVMPVLKS PTIPFFDPQI PKLTDLENLH LPLPLLQPLM QQVPQPIPQT LALPPQPLWSVPQPKVLPIP QQVVPYPQRA VPVQALLLNQ ELLLNPTHQI YPVTQPLAPV HNPISV
Greater than 95.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
CSN2 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.5M NaCl, 0.25M imidazole, 1mM DTT and 0.1mM PMSF.