missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CST3 Protein

A cDNA sequence encoding the CST3 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP10561-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15984248

  • 7540.00€ / 1mg
Expédition estimée: 17-01-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

CST3 Protein
Research Use Only
The inhibitory function of Cystatin-C on papin’s protease activity was measured by a colorimetric assay using L-BAPA as substrate. IC50 value was measured at 5-20 microg/ml (0.3-1.5 microm) with a range of 1.56-50microg/ml Cystatin-C in presence of 0.55microm papain and 0.44microm L-BAPA. The activity is typically 0.3-1.5microm IC50.
Human
Untagged
Greater than 95% as obsereved by SDS-PAGE.
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
Human (HEK293)
The Cystatin-C was lyophilized from 1 mg/ml in 1xPBS.
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions