missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CTLA 4 Protein
A cDNA sequence encoding the CTLA 4 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11555-25ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
1493 | |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSKAMHVAQ PAVVLASSRG IASFVCEYAS PGKATEVRVT VLRQADSQVT EVCAATYMMG NELTFLDDSI CTGTSSGNQV NLTIQGLRAM DTGLYICKVE LMYPPPYYLG IGNGTQIYVI DPEPCPDSD | |
Greater than 90% as determined by SDS-PAGE. |
CTLA 4 Protein | |
Research Use Only | |
CTLA4 | |
Human | |
His | |
The CTLA 4 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |