missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DCTN2 (1-401) Protein

A cDNA sequence encoding the DCTN2 (1-401) was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP14012-HIS-EC-20ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15975409

Quantité Prix
1 171.64€ / 20µg
10 161.27€ / 20µg
Date d’expédition estimée: 26-11-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!



Description

Description

Spécification

Spécification

DCTN2 (1-401) Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
DCTN2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl (pH 8.0), 20% glycerol, 0.15M NaCl and 1mM DTT.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMADPKYA DLPGIARNEP DVYETSDLPE DDQAEFDAFA QELEELTSTS VEHIIVNPNA AYDKFKDKRV GTKGLDFSDR IGKTKRTGYE SGEYEMLGEG LGVKETPQQK YQRLLHEVQE LTTEVEKIKT TVKESATEEK LTPVLLAKQL AALKQQLVAS HLEKLLGPDA AINLTDPDGA LAKRLLLQLE ATKNSKGGSG GKTTGTPPDS SLVTYELHSR PEQDKFSQAA KVAELEKRLT ELETAVRCDQ DAQNPLSAGL QGACLMETVE LLQAKVSALD LAVLDQVEAR LQSVLGKVNE IAKHKASVED ADTQSKVHQL YETIQRWSPI ASTLPELVQR LVTIKQLHEQ AMQFGQLLTH LDTTQQMIAN SLKDNTTLLT QVQTTMRENL ATVEGNFASI DERMKKLGK
Vidéos
FDS
Documents
Certificats