missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNAJB6 Protein
A cDNA sequence encoding the DNAJB6 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11677-5ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
DNAJB6 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMVDYYEV LGVQRHASPE DIKKAYRKLA LKWHPDKNPE NKEEAERKFK QVAEAYEVLS DAKKRDIYDK YGKEGLNGGG GGGSHFDSPF EFGFTFRNPD DVFREFFGGR DPFSFDFFED PFEDFFGNRR GPRGSRSRGT GSFFSAFSGF PSFGSGFSSF DTGFTSFGSL GHGGLTSFSS TSFGGSGMGN FKSISTSTKM VNGRKITTKR IVENGQERVE VEEDGQLKSL TINGVADDDA LAEERMRRGQ NALPAQPAGL RPPKPPRPAS LLRHAPHCLS EEEGEQDRPR APGPWDPLAS AAGLKEGGKR KKQKQREESK KKKSTKGNH | |
Greater than 85.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The DNAJB6 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 5mM DTT and 30% glycerol. |