Learn More
enQuireBio™ Recombinant Human DUSP14 / MKP-6 Protein
A DNA sequence encoding the Homo sapiens (Human) DUSP14 / MKP-6, was expressed in the hosts and tags indicated.
Marque:  enQuireBio™ QP5957-ec-100ug
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15968929
Quantité Prix
1 434.28€ / 100µg
6 408.22€ / 100µg
Date d’expédition estimée: 18-11-2021
Spécification
DUSP14 / MKP-6 Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
DUSP14
Recombinant Protein
E. coli
MSSRGHSTLP RTLMAPRMIS EGDIGGIAQI TSSLFLGRGS VASNRHLLQA RGITCIVNAT IEIPNFNWPQ FEYVKVPLAD MPHAPIGLYF DTVADKIHSV SRKHGATLVH CAAGVSRSAT LCIAYLMKFH NVCLLEAYNW VKARRPVIRP NVGFWRQLID YERQLFGKST VKMVQTPYGI VPDVYEKESR HLMPYWGI