missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EDF1 Protein
A cDNA sequence encoding the EDF1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11745-25ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
EDF1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAESDWDTVT VLRKKGPTAA QAKSKQAILA AQRRGEDVET SKKWAAGQNK QHSITKNTAK LDRETEELHH DRVTLEVGKV IQQGRQSKGL TQKDLATKIN EKPQVIADYE SGRAIPNNQV LGKIERAIGL KLRGKDIGKP IEKGPRAKLE HHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
0.5 mg/ml solution containing 20mM Tris pH 8.0, 1mM DTT, 0.1M NaCl and 20% glycerol. |