missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Eyes absent homolog 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Eyes absent homolog 2, was expressed in the hosts and tags indicated.

Marque:  enQuireBio™ QP6005-ec-10ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15909299

Cet article est actuellement indisponible ou discontinu. Veuillez consulter la page produit pour de possible alternative dans l’onglet


Description

Description

Spécification

Spécification

NP_005235.3
Eyes absent homolog 2 Protein
Research Use Only
EYA2
Recombinant Protein
E. coli
MVELVISPSL TVNSDCLDKL KFNRADAAVW TLSDRQGITK SAPLRVSQLF SRSCPRVLPR QPSTAMAAYG QTQYSAGIQQ ATPYTAYPPP AQAYGIPSFS TSPTGQSPYT YQMHGTTGFY QGGNGLGNAA GFGSVHQDYP SYPGFPQSQY PQYYGSSYNP PYVPASSICP SPLSTSTYVL QEASHNVPNQ SSESLAGEYN THNGPSTPAK EGDTDRPHRA SDGKLRGRSK RSSDPSPAGD NEIERVFVWD LDETIIIFHS LLTGTFASRY GKDTTTSVRI GLMMEEMIFN LADTHLFFND LEDCDQIHVD DVSSDDNGQD LSTYNFSADG FHSSAPGANL CLGSGVHGGV DWMRKLAFRY RRVKEMYNTY KNNVGGLIGT PKRETWLQLR AELEALTDLW LTHSLKALNL INSRPNCVNV LVTTTQLIPA LAKVLLYGLG SVFPIENIYS ATKTGKESCF ERIMQRFGRK AVYVVIGDGV EEEQGAKKHN MPFWRISCHA DLEALRHALE LEYL
Greater than 90% as determined by SDS-PAGE.
2139
10 μg
Not determined.
Partial (see sequence information for more details). of Isoform 2
Human
His-SUMO
Tris-based buffer, 50% glycerol
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions