missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GAPDH Protein

A DNA sequence encoding the Homo sapiens (Human) GAPDH, was expressed in the hosts and tags indicated.

Marque:  enQuireBio™ QP6070-ec-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15969759

Quantité Prix
1 1979.08€ / 1mg
3 1861.92€ / 1mg
Date d’expédition estimée: 22-11-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!



Description

Description

Spécification

Spécification

NP_001243728.1
GAPDH Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
2597
Greater than 90% as determined by SDS-PAGE.
Research Use Only
GAPDH
Recombinant Protein
E. coli
GKVKVGVNGF GRIGRLVTRA AFNSGKVDIV AINDPFIDLN YMVYMFQYDS THGKFHGTVK AENGKLVING NPITIFQERD PSKIKWGDAG AEYVVESTGV FTTMEKAGAH LQGGAKRVII SAPSADAPMF VMGVNHEKYD NSLKIISNAS CTTNCLAPLA KVIHDNFGIV EGLMTTVHAI TATQKTVDGP SGKLWRDGRG ALQNIIPAST GAAKAVGKVI PELNGKLTGM AFRVPTANVS VVDLTCRLEK PAKYDDIKKV VKQASEGPLK GILGYTEHQV VSSDFNSDTH SSTFDAGAGI ALNDHFVKLI SWYDNEFGYS NRVVDLMAHM ASKE
Vidéos
FDS
Documents
Certificats