missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GHRH Protein

A cDNA sequence encoding the GHRH was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP10643-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15974498

  • 203.00€ / 1mg
Expédition estimée: 28-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

GHRH Protein
Research Use Only
GHRH increases plasma growth hormone concentrations by directly stimulating the anterior pituitary gland to release natural human growth hormone.
Human
Untagged
The GHRH peptide (1 mg/ml) was lyophilized after extensive dialyses against 1.7 mg sodium phosphate buffer (0.1 mg sodium phosphate monobasic and 1.6 mg sodium phosphate dibasic).
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
The free base of sermorelin has the empirical formula C149H246N 44O42STyr-Ala-Asp-Ala-Ile-Phe-Thr-Asn-Ser-Tyr-Arg-Lys-Val-Leu-Gly-Gln-Leu-Ser-Ala-Arg-Lys-Leu-Leu-Gln-Asp-Ile-Met-Ser-Arg-NH2
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions