Learn More
enQuireBio™ Recombinant Human GM-CSF / CSF2 Protein
Recombinant Protein
Marque:  enQuireBio™ QP5231-500ug
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15925749
  • 2585.00€ / 500µg
Expédition estimée: 10-05-2024
Spécification
NP_000749.2
GM-CSF / CSF2 Protein
Research Use Only
CSF2
Recombinant Protein
E. coli
MAPARSPSPS TQPWEHVNAI QEARRLLNLS RDTAAEMNET VEVISEMFDL QEPTCLQTRL ELYKQGLRGS LTKLKGPLTM MASHYKQHCP PTPETSCATQ IITFESFKEN LKDFLLVIPF DCWEPVQE
Greater than 95% as determined by SDS-PAGE.
1437
500 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity at a concentration of 200 pg/mL in the TF-1 cell proliferation assay. The bioactivity acceptance criteria is set at 5 x 10^6 units/mg.
Human
Untagged
10 mM sodium phosphate,pH 7.5