missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HNRNPAB Protein

A cDNA sequence encoding the HNRNPAB was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP12290-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15999948

  • 7540.00€ / 1mg
Expédition estimée: 10-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

HNRNPAB Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSEAGEE QPMETTGATE NGHEAVPEGE SPAGAGTGAA AGAGGATAAP PSGNQNGAEG DQINASKNEE DAGKMFVGGL SWDTSKKDLK DYFTKFGEVV DCTIKMDPNT GRSRGFGFIL FKDAASVEKV LDQKEHRLDG RVIDPKKAMA MKKDPVKKIF VGGLNPEATE EKIREYFGEF GEIEAIELPM DPKLNKRRGF VFITFKEEEP VKKVLEKKFH TVSGSKCEIK VAQPKEVYQQ QQYGSGGRGN RNRGNRGSGG GGGGGGQGST NYGKSQRRGG HQNNYKPY
Greater than 95.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
HNRNPAB protein solution (0.25 mg/ml) containing 30% glycerol and 1mM DTT.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions