missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HSBP1L1 Protein

A cDNA sequence encoding the HSBP1L1 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP12318-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15980039

  • 3740.00€ / 1mg
Expédition estimée: 12-07-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

HSBP1L1 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
HSBP1L1 protein solution (1 mg/ml) containing Phosphate buffer saline (pH 7.4) and 10% glycerol.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMDVRGPE APGGRALRDA AENLFQELQE HFQALTATLN LRMEEMGNRI EDLQKNVNDL MVQAGIENSI KEQMLKT
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions