missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HSFY1 Protein

A cDNA sequence encoding the HSFY1 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP12324-5ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15980059

  • 125.00€ / 5µg
Expédition estimée: 10-07-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

HSFY1 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The HSFY1 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea.
Greater than 80.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAHVSSE TQDVSPKDEL TASEASTRSP LCEHTFPGDS DLRSMIEEHA FQVLSQGSLL ESPSYTVCVS EPDKDDDFLS LNFPRKLWKI VESDQFKSIS WDENGTCIVI NEELFKKEIL ETKAPYRIFQ TDAIKSFVRQ LNLYGFSKIQ QNFQRSAFLA TFLSEEKESS VLSKLKFYYN PNFKRGYPQL LVRVKRRIGV KNASPISTLF NEDFNKKHFR AGANMENHNS ALAAEASEES LFSASKNLNM PLTRESSVRQ IIANSSVPIR SGFPPPSPST SVGPSEQIAT DQHAILNQLT TIHMHSHSTY MQARGHIVNF ITTTTSQYHI ISPLQNGYFG LTVEPSAVPT RYPLVSVNEA PYRNMLPAGN PWLQMPTIAD RSAAPHSRLA LQPSPLDKYH PNYN
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions