missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL 28B Protein
A cDNA sequence encoding the IL 28B was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10709-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
282617 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The specific activity was determined by the dose dependent protection of the cytopathic effect on A549 cells (human lung adenocarcinoma epithelial cell line) that were challenged with encephalomyocarditis (EMC) virus and is typically 0.1-1ng/ml. | |
Human | |
Untagged | |
The IL28B was lyophilized from 1 mg/ml in 1xPBS. |
IL 28B Protein | |
Research Use Only | |
IFNL3 | |
Recombinant Protein | |
Human (HEK293) | |
MTGDCMPVLV LMAAVLTVTG AVPVARLRGA LPDARGCHIA QFKSLSPQEL QAFKRAKDAL EESLLLKDCK CRSRLFPRTW DLRQLQVRER PVALEAELAL TLKVLEATAD TDPALGDVLD QPLHTLHHIL SQLRACIQPQ PTAGPRTRGR LHHWLHRLQE APKKESPGCL EASVTFNLFR LLTRDLNCVA SGDLCV | |
Greater than 95% as obsereved by SDS-PAGE. |