Learn More
enQuireBio™ Recombinant Human KIR3DL1 Protein
A cDNA sequence encoding the KIR3DL1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12492-5ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
KIR3DL1 Protein | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The protein (1 mg/ml) contains 25mM Tris-HCl (pH 7.5) and 100mM NaCl. |
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSTSGT IDKLDIEFHLWCSNKKNAAV MDQEPAGNRT ANSEDSDEQD PEEVTYAQLD HCVFTQRKIT RPSQRPKTPP TDTILYTELP NAKPRSKVVS CP |