Learn More
enQuireBio™ Recombinant Human KLF7 Protein
A cDNA sequence encoding the KLF7 was constructed and used to recombinantly synthesize the protein.
Marque:  enQuireBio™ QP12500-1mg
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15920619
  • 3915.00€ / 1mg
Expédition estimée: 10-05-2024
Spécification
KLF7 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMDVLASY SIFQELQLVH DTGYFSALPS LEETWQQTCL ELERYLQTEP RRISETFGED LDCFLHASPP PCIEESFRRL DPLLLPVEAA ICEKSSAVDI LLSRDKLLSE TCLSLQPASS SLDSYTAVNQ AQLNAVTSLT PPSSPELSRH LVKTSQTLSA VDGTVTLKLV AKKAALSSVK VGGVATAAAA VTAAGAVKSG QSDSDQGGLG AEACPENKKR VHRCQFNGCR KVYTKSSHLK AHQRTHTGEK PYKCSWEGCE WRFARSDELT RHYRKHTGAK PFKCNHCDRC FSRSDHLALH MKRHI
Greater than 90% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The KLF7 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol.