Learn More
enQuireBio™ Recombinant Human Latexin Protein
A cDNA sequence encoding the Latexin was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12541-5ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
Latexin Protein | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The Latexin protein solution contains 20mM Tris-HCl, pH 7.5, 50mM NaCl and 10% Glycerol. |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MEIPPTNYPA SRAALVAQNY INYQQGTPHR VFEVQKVKQA SMEDIPGRGH KYRLKFAVEE IIQKQVKVNC TAEVLYPSTG QETAPEVNFTFEGETGKNPD EEDNTFYQRL KSMKEPLEAQ NIPDNFGNVS PEMTLVLHLA WVACGYIIWQ NSTEDTWYKM VKIQTVKQVQ RNDDFIELDYTILLHNIASQ EIIPWQMQVL WHPQYGTKVK HNSRLPKEVQ LE |