missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LBP Protein

A cDNA sequence encoding the LBP was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP10741-5ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15975028

Quantité Prix
1 343.29€ / 5µg
6 322.56€ / 5µg
Date d’expédition estimée: 23-11-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!



Description

Description

Spécification

Spécification

3929
5 μg
< 1.0 EU per ug protein as determined by the LAL method.
Up to 0.2 microg/ ml LBP mediates binding of FITC-LPS (0.5microg/ml) to CD14+CHO transfectants (FACS).
Human
His
Recombinant Human LBP was lyophilized from a protein solution (0.3 mg/ml) containing phosphate-buffered saline, pH 7.2.
LBP Protein
Research Use Only
LBP
Recombinant Protein
CHO
MGALARALPS ILLALLLTST PEALGANPGL VARITDKGLQ YAAQEGLLAL QSELLRITLP DFTGDLRIPH VGRGRYEFHS LNIHSCELLH SALRPVPGQG LSLSISDSSI RVQGRWKVRK SFFKLQGSFD VSVKGISISV NLLLGSESSG RPTVTASSCS SDIADVEVDM SGDLGWLLNL FHNQIESKFQ KVLESRICEM IQKSVSSDLQ PYLQTLPVTT EIDSFADIDY SLVEAPRATA QMLEVMFKGE IFHRNHRSPV TLLAAVMSLP EEHNKMVYFA ISDYVFNTAS LVYHEEGYLN FSITDDMIPP DSNIRLTTKS FRPFVPRLAR LYPNMNLELQ GSVPSAPLLN FSPGNLSVDP YMEIDAFVLL PSSSKEPVFR LSVATNVSAT LTFNTSKITG FLKPGKVKVE LKESKVGLFN AELLEALLNY YILNTFYPKF NDKLAEGFPL PLLKRVQLYD LGLQIHKDFL FLGANVQYMR V
Vidéos
FDS
Documents
Certificats