missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MBD3 Protein

A cDNA sequence encoding the MBD3 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP12654-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15981119

  • 7540.00€ / 1mg
Expédition estimée: 19-01-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

MBD3 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMERKRWE CPALPQGWER EEVPRRSGLS AGHRDVFYYS PSGKKFRSKP QLARYLGGSM DLSTFDFRTG KMLMSKMNKS RQRVRYDSSN QVKGKPDLNT ALPVRQTASI FKQPVTKITN HPSNKVKSDP QKAVDQPRQL FWEKKLSGLN AFDIAEELVK TMDLPKGLQG VGPGCTDETL LSAIASALHT STMPITGQLS AAVEKNPGVW LNTTQPLCKA FMVTDEDIRK QEELVQQVRK RLEEALMADM LAHVEELARD GEAPLDKACA EDDDEEDEEE EEEEPDPDPE MEHV
Greater than 85% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The MBD3 solution (0.25 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 1mM DTT, 1mM EDTA and 40% glycerol.
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions