missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NNT1 Protein

A cDNA sequence encoding the NNT1 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP10794-10ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15965188

  • 203.00€ / 10µg
Expédition estimée: 10-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

23529
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
The ED50, as measured in a cell proliferation assay using human TF?1 cells transfected with human CNTF R?, is less than 15ng/ml.
Human
Untagged
NNT-1 protein was lyophilized from a 0.2microm filtered solution in Acetonitrile and TFA.
NNT1 Protein
Research Use Only
CLCF1
Recombinant Protein
E. coli
MLNRTGDPGP GPSIQKTYDL TRYLEHQLRS LAGTYLNYLG PPFNEPDFNP PRLGAETLPR ATVDLEVWRS LNDKLRLTQN YEAYSHLLCY LRGLNRQAAT AELRRSLAHF CTSLQGLLGS IAGVMAALGY PLPQPLPGTE PTWTPGPAHS DFLQKMDDFW LLKELQTWLW RSAKDFNRLK KKMQPPAAAV TLHLGAHGF
Greater than 95.0% as determined by SDS-PAGE.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions