missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NPPA Protein

A cDNA sequence encoding the NPPA was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP12873-10ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15971809

  • 203.00€ / 10µg
Expédition estimée: 10-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

4878
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MKHHHHHHNP MYNAVSNADL MDFKNLLDHL EEKMPLEDEV VPPQVLSEPN EEAGAALSPL PEVPPWTGEV SPAQRDGGAL GRGPWDSSDR SALLKSKLRA LLTAPR
Greater than 95.0% as determined by SDS-PAGE.
NPPA Protein
Research Use Only
NPPA
Human
His
NPPA filtered (0.4 microm) and lyophilized from 0.5 mg/ml in 20 mM Tris buffer, 50 mM NaCl and 5% w/v trehalosa, pH 7.5.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions