missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NRG1 A Protein

A cDNA sequence encoding the NRG1 A was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP10797-5ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15975198

Quantité Prix
1 114.77€ / 5µg
10 107.33€ / 5µg
Date d’expédition estimée: 26-11-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!



Description

Description

Spécification

Spécification

NRG1 A Protein
5 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
SHLVKCAEKE KTFCVNGGEC FMVKDLSNPS RYLCKCQPGF TGARCTENVP MKVQNQEKAE ELYQK
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
The ED50 was determined by the dose-dependent stimulation of the proliferation of human MCF-7 cells is less than 40 ng/ml, corresponding to a specific activity of > 2.5M 104 units/mg.
Human
Untagged
Lyophilized from a 0.2microm filtered solution in 20mM PBS, pH 6.0 and 150mM NaCl.
Vidéos
FDS
Documents
Certificats