Learn More
enQuireBio™ Recombinant Human NRG1 B1 Protein
A cDNA sequence encoding the NRG1 B1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10798-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NRG1 B1 Protein | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
SHLVKCAEKE KTFCVNGGEC FMVKDLSNPS RYLCKCPNEF TGDRCQNYVM ASFYKHLGIE FMEAE |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. | |
Research Use Only | |
The ED50 was determined by the dose-dependent stimulation of the proliferation of human MCF-7 cells is less than 0.5ng/ml, corresponding to a specific activity of >2.0M 106 units/mg. | |
Human | |
Untagged | |
Lyophilized from a 0.2microm filtered solution in 1M PBS, pH 7.4 and 5% trehalose. |