missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NRG1-beta 1A Protein
Recombinant Protein
Marque: enQuireBio™ QP5212-100ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_039250.2 | |
100 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MSHLVKCAEK EKTFCVNGGE CFMVKDLSNP SRYLCKCPNE FTGDRCQNYV MASFYKHLGI EFMEAE | |
Greater than 95% as determined by SDS-PAGE. |
NRG1-beta 1A Protein | |
Research Use Only | |
NRG1 | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |