missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human OLFM1 Protein
A cDNA sequence encoding the OLFM1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12923-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
OLFM1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSLPTNPEE SWQVYSSAQD SEGRCICTVV APQQTMCSRD ARTKQLRQLL EKVQNMSQSI EVLDRRTQRD LQYVEKMENQ MKGLESKFKQ VEESHKQHLA RQFKG | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
OLFM1 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer, (pH 7.5), 0.2M NaCl, 50% glycerol and 5mM DTT. |