missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human OSR2 Protein

A cDNA sequence encoding the OSR2 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP12934-5ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15981999

  • 131.00€ / 5µg
Expédition estimée: 19-01-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

OSR2 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMGSKALP APIPLHPSLQ LTNYSFLQAV NTFPATVDHL QGLYGLSAVQ TMHMNHWTLG YPNVHEITRS TITEMAAAQG LVDARFPFPA LPFTTHLFHP KQGAIAHVLP ALHKDRPRFD FANLAVAATQ EDPPKMGDLS KLSPGLGSPI SGLSKLTPDR KPSRGRLPSK TKKEFICKFC GRHFTKSYNL LIHERTHTDE RPYTCDICHK AFRRQDHLRD HRYIHSKEKP FKCQECGKGF CQSRTLAVHK TLHMQTSSPT AASSAAKCSG ETVICGGTA
Greater than 80% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
OSR2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea.
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions