missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PINK1 Protein
A cDNA sequence encoding the PINK1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13056-20ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
PINK1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MYLIGQSIGK GCSAAVYEAT MPTLPQNLEV TKSTGLLPGR GPGTSAPGEG QERAPGAPAF PLAIKMMWNI SAGSSSEAIL NTMSQELVPA SRVALAGEYG AVTYRKSKRG PKQLAPHPNI IRVLRAFTSS VPLLPGALVD YPDVLPSRLH PEGLGHGRTL FLVMKNYPCT LRQYLCVNTP SPRLAAMMLL QLLEGVDHLV QQGIAHRDLK SDNILVELDP DGCPWLVIAD FGCCLADESI GLQLPFSSWY VDRGGNGCLM APEVSTARPG PRAVIDYSKA DAWAVGAIAY EIFGLVNPFY GQGKAHLESR SYQEAQLPAL PESVPPDVRQ LVRALLQREA SKRPSARVAA NVL | |
Greater than 90% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The PINK1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1M Urea and 5% glycerol. |