Learn More
enQuireBio™ Recombinant Human PNPO Protein
A cDNA sequence encoding the PNPO was constructed and used to recombinantly synthesize the protein.
Marque:  enQuireBio™ QP13085-20ug
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15972459
  • 203.00€ / 20µg
Expédition estimée: 10-05-2024
Spécification
PNPO Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MDPVKQFAAW FEEAVQCPDI GEANAMCLAT CTRDGKPSAR MLLLKGFGKD GFRFFTNFES RKGKELDSNP FASLVFYWEP LNRQVRVEGP VKKLPEEEAE CYFHSRPKSS QIGAVVSHQS SVIPDREYLR KKNEELEQLY QDQEVPKPKS WGGYVLYPQV MEFWQGQTNR LHDRIVFRRG LPTGDSPLGP MTHRGEEDWL YERLAP
Greater than 90.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The PNPO solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.1M NaCl and 0.1mM PMSF.