missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human POLR2K Protein
A cDNA sequence encoding the POLR2K was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13105-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
POLR2K Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDTQKDV QPPKQQPMIY ICGECHTENE IKSRDPIRCR ECGYRIMYKK RTKRLVVFDA R | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
POLR2K protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |