Learn More
enQuireBio™ Recombinant Human Potassium-transporting ATPase subunit beta Protein
A DNA sequence encoding the Homo sapiens (Human) Potassium-transporting ATPase subunit beta, was expressed in the hosts and tags indicated.
Marque:  enQuireBio™ QP5692-ye-500ug
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15967049
  • 1665.00€ / 500µg
Expédition estimée: 10-05-2024
Spécification
NP_000696.1
Potassium-transporting ATPase subunit beta Protein
Research Use Only
ATP4B
Recombinant Protein
Yeast
CLYVLMQTVD PYTPDYQDQL RSPGVTLRPD VYGEKGLEIV YNVSDNRTWA DLTQTLHAFL AGYSPAAQED SINCTSEQYF FQESFRAPNH TKFSCKFTAD MLQNCSGLAD PNFGFEEGKP CFIIKMNRIV KFLPSNGSAP RVDCAFLDQP RELGQPLQVK YYPPNGTFSL HYFPYYGKKA QPHYSNPLVA AKLLNIPRNA EVAIVCKVMA EHVTFNNPHD PYEGKVEFKL KIEK
Greater than 90% as determined by SDS-PAGE.
496
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol