Learn More
enQuireBio™ Recombinant Human PPIF Protein
A cDNA sequence encoding the PPIF was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10829-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
10105 | |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
PPIF | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH CSKGSGDPSS SSSSGNPLVY LDVDANGKPL GRVVLELKAD VVPKTAENFR ALCTGEKGFG YKGSTFHRVI PSFMCQAGDF TNHNGTGGKS IYGSRFPDEN FTLKHVGPGV LSMANAGPNT NGSQFFICTI KTDWLDGKHV VFGHVKEGMD VVKKIESFGS KSGRTSKKIV ITDCGQLS |
PPIF Protein | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 250 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-Hcl pH8.0 using chymotrypsin. | |
Human | |
His | |
PPIF solution containing 20mM Tris-HCl pH 7.5, 10% glycerol and 1mM DTT. |