Learn More
enQuireBio™ Recombinant Human PRKCE Protein
A cDNA sequence encoding the PRKCE was constructed and used to recombinantly synthesize the protein.
Marque:  enQuireBio™ QP13145-10ug
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15972639
Quantité Prix
1 171.64€ / 10µg
10 161.27€ / 10µg
Date d’expédition estimée: 22-11-2021
Spécification
PRKCE Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
PRKCE is supplied in 20mM Tris, pH 7.4, 0.1M NaCl, 20% glycerol, 1mM DTT, 0.1mM EDTA, 0.2mM PMSF, 1mM benzamidine and 0.03% Brij-35.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
MVVFNGLLKI KICEAVSLKP TAWSLRHAVG PRPQTFLLDP YIALNVDDSR IGQTATKQKT NSPAWHDEFV TDVCNGRKIE LAVFHDAPIG YDDFVANCTI QFEELLQNGS RHFEDWIDLE PEGRVYVIID LSGSSGEAPK DNEERVFRER MRPRKRQGAV RRRVHQVNGH KFMATYLRQP TYCSHCRDFI WGVIGKQGYQ CQVCTCVVHK RCHELIITKC AGLKKQETPD QVGSQRFSVN MPHKFGIHNY KVPTFCDHCG SLLWGLLRQG LQCKVCKMNV HRRCETNVAP NCGVDARGIA KVLADLGVTP DKITNSGQRR KKLIAGAESP QPASGSSPSE EDRSKSAPTS PCDQEIKELE NNIRKALSFD NRGEEHRAAS SPDGQLMSPG ENGEVRQGQA KRLGLDEFNF IKVLGKGSFG KVMLAELKGK DEVYAVKVLK KDVILQDDDV DCTMTEKRIL ALARKHPYLT QLYCCFQTKD RLFFVMEYVN GGDLMFQIQR SRKFDEPRSR FYAAEVTSAL MFLHQHGVIY RDLKLDNILL DAEGHCKLAD FGMCKEGILN GVTTTTFCGT PDYIAPEILQ ELEYGPSVDW WALGVLMYEM MAGQPPFEAD NEDDLFESIL HDDVLYPVWL SKEAVSILKA FMTKNPHKRL GCVASQNGED AIKQHPFFKE IDWVLLEQKK IKPPFKPRIK TKRDVNNFDQ DFTREEPVLT LVDEAIVKQI NQEEFKGFSY FGEDLMP