missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Probable G-protein coupled receptor 75 Protein
A DNA sequence encoding the Homo sapiens (Human) Probable G-protein coupled receptor 75, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP6113-ye-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_006785.1 | |
Probable G-protein coupled receptor 75 Protein | |
Research Use Only | |
GPR75 | |
Recombinant Protein | |
Yeast | |
NPFIYSRNSA GLRRKVLWCL QYIGLGFFCC KQKTRLRAMG KGNLEVNRNK SSHHETNSAY MLSPKPQKKF VDQACGPSHS KESMVSPKIS AGHQHCGQSS STPINTRIEP YYSIYNSSPS QEESSPCNLQ PVNSFGFANS YIAMHYHTTN DLVQEYDSTS AKQIPVPSV | |
Greater than 90% as determined by SDS-PAGE. |
10936 | |
10 μg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |