missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RBP5 Protein
A cDNA sequence encoding the RBP5 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13284-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
RBP5 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPPNLTGYYR FVSQKNMEDY LQALNISLAV RKIALLLKPD KEIEHQGNHM TVRTLSTFRN YTVQFDVGVE FEEDLRSVDG RKCQTIVTWE EEHLVCVQKG EVPNRGWRHW LEGEMLYLEL TARDAVCEQV FRKVR | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RBP5 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 2mM DTT and 100mM NaCl. |