missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RPL22 Protein

A cDNA sequence encoding the RPL22 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP13343-2ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15973269

Quantité Prix
1 114.77€ / 2µg
10 107.33€ / 2µg
Date d’expédition estimée: 18-11-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!



Description

Description

Spécification

Spécification

RPL22 Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The RPL22 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 20% glycerol.
Greater than 85% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAPVKKL VVKGGKKKKQ VLKFTLDCTH PVEDGIMDAA NFEQFLQERI KVNGKAGNLG GGVVTIERSK SKITVTSEVP FSKRYLKYLT KKYLKKNNLR DWLRVVANSK ESYELRYFQI NQDEEEEEDE D
Vidéos
FDS
Documents
Certificats