missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RPL34 Protein
A cDNA sequence encoding the RPL34 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13347-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
RPL34 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMVQRLTY RRRLSYNTAS NKTRLSRTPG NRIVYLYTKK VGKAPKSACG VCPGRLRGVR AVRPKVLMRL SKTKKHVSRA YGGSMCAKCV RDRIKRAFLI EEQKIVVKVL KAQAQSQKAK | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RPL34 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |