Learn More
enQuireBio™ Recombinant Human RPS4X Protein
A cDNA sequence encoding the RPS4X was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13358-5ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
RPS4X Protein | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RPS4X solution (0.5 mg/ml) contains 20mM Tris-HCl(pH 8.0), 40% glycerol, 0.15M NaCl and 1mM DTT. |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMARGPKK HLKRVAAPKH WMLDKLTGVF APRPSTGPHK LRECLPLIIF LRNRLKYALT GDEVKKICMQ RFIKIDGKVR TDITYPAGFM DVISIDKTGE NFRLIYDTKG RFAVHRITPE EAKYKLCKVR KIFVGTKGIP HLVTHDARTI RYPDPLIKVN DTIQIDLETG KITDFIKFDT GNLCMVTGGA NLGRIGVITN RERHPGSFDV VHVKDANGNS FATRLSNIFV IGKGNKPWIS LPRGKGIRLT IAEERDKRLA AKQSSG |